Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109973 Supplier Invitrogen™ Supplier No. PA5109973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLALRRPEIL VQPLKVSNRK SSGFRKSFRL SRKDKKTNKS MYECKKSDQY DTADVPTYEE VTPYRRQTNE KYRLVVLVGP VGVGLNELKR KLLISDTQHY GVTVPHTTRA RRSQESDGVE YIFISKHLFE TDVQNNKFIE YGEYKNNYYG TSIDSVRSVL AKNKVCLLDV QPHTVKHLRT LEFKPYVIFI KPPSIERLRE TRKNAKIISS RDDQGAAKPF TEEDFQEMIK SAQIMESQYG HLFDKIIIND DLTVAFNELK TTFDKLETET HWVPVSWLHS.

Acts as an important adapter that promotes epithelial cell polarity and tight junction formation via its interaction with DLG1. Involved in the assembly of protein complexes at sites of cell-cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MPP7
Gene Accession No. Q8BVD5
Gene Alias 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MAGUK p55 subfamily member 7; membrane palmitoylated protein 7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7); MPP7
Gene Symbols MPP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 297-576 of human MPP7 (NP_775767.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75739
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.