Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mpp8 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171809FR
Description
mpp8 Polyclonal antibody specifically detects mpp8 in Human samples. It is validated for Western BlotSpecifications
mpp8 | |
Polyclonal | |
Western Blot | |
FLJ35237, HSMPP8, M-phase phosphoprotein 8, M-phase phosphoprotein, mpp, M-phase phosphoprotein, mpp8, MPP8, TWA3, Two hybrid-associated protein 3 with RanBPM | |
The immunogen for this product maps to a region between residue 200 and 250 of human M-phase Phosphoprotein 8 using the numbering given in entry NP_059990.2 (GeneID 54737). | |
0.1 mL | |
Cell Cycle and Replication | |
54737 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction