Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mpp8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171808
Description
mpp8 Polyclonal specifically detects mpp8 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
mpp8 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FLJ35237, HSMPP8, M-phase phosphoprotein 8, M-phase phosphoprotein, mpp, M-phase phosphoprotein, mpp8, MPP8, TWA3, Two hybrid-associated protein 3 with RanBPM | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
NP_059990.2 | |
MPHOSPH8 | |
The immunogen for this product maps to a region between residue 1 and 50 of human M-phase Phosphoprotein 8 using the numbering given in entry NP_059990.2 (GeneID 54737). | |
100 μL | |
Cell Cycle and Replication | |
54737 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction