Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MPP8 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-560-479
![]() ![]() |
Proteintech Group Inc
167961AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Heterochromatin component that specifically recognizes and binds methylated ′Lys-9′ of histone H3 (H3K9me) and promotes recruitment of proteins that mediate epigenetic repression. Mediates recruitment of the HUSH complex to H3K9me3 sites: the HUSH complex is recruited to genomic loci rich in H3K9me3 and is probably required to maintain transcriptional silencing by promoting recruitment of SETDB1, a histone methyltransferase that mediates further deposition of H3K9me3. Binds H3K9me and promotes DNA methylation by recruiting DNMT3A to target CpG sites; these can be situated within the coding region of the gene. Mediates down-regulation of CDH1 expression.Specifications
MPP8 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
G3V8T1, Q3TYA6, Q99549 | |
290270, 54737, 75339 | |
MPP8 Fusion Protein Ag10268 | |
Primary | |
-20°C | |
MPHOSPH8 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HSMPP8, M phase phosphoprotein 8, MPHOSPH8, MPP8, TWA3 | |
Mphosph8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title