Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536981 Supplier Invitrogen™ Supplier No. PA536981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 78 and 116 kDa. Purity is >95% by SDS-PAGE.

Progression of cells from interphase to mitosis involves alterations in cell structures and activities. The transition from G2 to M phase is induced by M phase-promoting factor (MPF). In M phase, many proteins are phosphorylated directly by MPF or indirectly by kinases activated by MPF. These M phase phosphoproteins (MPPs), also known as MPHOSPHs, permit disassembly of interphase structures and generation of M phase enzymatic activities and structures. MPP9 (M-phase phosphoprotein 9), also known as MPHOSPH9, is a 1,031 amino acid peripheral membrane protein of the Golgi apparatus that exists as two alternatively spliced isoforms. The gene encoding MPP9 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP9
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MPHOSPH9
Gene Accession No. A6H5Y1, Q99550
Gene Alias 4930548D04Rik; 9630025B04Rik; AW413446; AW547060; B930097C17Rik; C87456; M-phase phosphoprotein 9; MPHOSPH9; MPP9; MPP-9
Gene Symbols MPHOSPH9
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 81-126 of human MPP9.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10198, 269702, 288654
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.