Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPPE1 Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA524488
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524488 Supplier Invitrogen™ Supplier No. MA524488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LLFCEFLIYY LAIFQCNWPE VKTTASDGEQ TTREPVLKAM FLADTHLLGE FLGHWLDKLR REWQMERAFQ TALWLLQPEV VFILGDIFDE GKWSTPEAWA DDVERFQKMF RHPSHVQLKV VAGNHDIGFH YEMNTYKVER FEKVFSSERL FSWKGINFVM VNSVALNGDG CGICSETEAE LIEVSHRLNC SRELLWWLQP RLVLSGHTHS ACEVHHGGRV PELSVPSFSW RNRNNPSFIM GSITPTDYTL SKCYLPREDV VLIIYCGVVG FLVVLTLTHF GLLASPFLSG LNLLGKRKTR.

Metallophosphoesterase required for transport of GPI-anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPPE1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E4
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MPPE1
Gene Accession No. Q53F39
Gene Alias A530095G11; metallo phosphoesterase; metallophosphoesterase 1; Mppe1; PGAP5; post-GPI attachment to proteins factor 5; PP579
Gene Symbols MPPE1
Host Species Mouse
Immunogen MPPE1 (AAH02877.1, 34 a.a. ~ 333 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65258
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.