Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPPE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552965 Supplier Invitrogen™ Supplier No. PA552965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEVKTTASDG EQTTREPVLK AMFLADTHLL GEFLGHWLDK LRREWQMERA FQTALWLLQP EVVFILGDIF DEGKWSTPEA WADDVERFQK MFRHPSHVQL KVVAGNHDIG FHYEMN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

Metallophosphoesterase required for transport of GPI-anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPPE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MPPE1
Gene Accession No. Q53F39
Gene Alias A530095G11; metallo phosphoesterase; metallophosphoesterase 1; Mppe1; PGAP5; post-GPI attachment to proteins factor 5; PP579
Gene Symbols MPPE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPPE1. Recombinant protein control fragment (Product #RP-88831).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.