Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPPE1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549047
Description
Predicted to react with monkey based on sequence homology.
Metallophosphoesterase required for transport of GPI- anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins.
Specifications
MPPE1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q53F39 | |
MPPE1 | |
KLH conjugated synthetic peptide between 261-289 amino acids from the C-terminal region of human MPPE1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MPPE1 | |
Metallophosphoesterase 1, 31--, Post-GPI attachment to proteins factor 5, MPPE1, PGAP5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65258 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction