Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPPED2/Brain protein 239 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1756 Supplier Bioss Supplier No. BS11888R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.

Specifications

Antigen MPPED2/Brain protein 239
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MPPED2
Gene Accession No. Q15777
Gene Alias 239FB; 2700082O15Rik; AV354767; AW060960; BOS_15029; brain protein 239-like; brpl; C11orf8; C11orf8h; cb1097; FAM1B; Fetal brain protein 239; Fetal brain protein 239 homolog; fk34e08; LOC100357974; metallophosphoesterase domain containing 2; metallophosphoesterase domain containing 2b; Metallophosphoesterase domain-containing protein 2; Metallophosphoesterase MPPED2; MPPED2; putative C11orf8 homolog; wu:fk34e08; zgc:56667
Gene Symbols MPPED2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MPPED2/Brain protein 239.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 744
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.