Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPPED2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112916
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112916 Supplier Invitrogen™ Supplier No. PA5112916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MPPED2 Polyclonal Antibody for Western Blot, ELISA

Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPPED2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MPPED2
Gene Accession No. Q15777, Q9CZJ0
Gene Alias 239FB; 2700082O15Rik; AV354767; AW060960; brain protein 239-like; brpl; C11orf8; C11orf8h; cb1097; FAM1B; Fetal brain protein 239; fetal brain protein 239 homolog; fk34e08; metallophosphoesterase domain containing 2; metallophosphoesterase domain containing 2b; metallophosphoesterase domain-containing protein 2; Metallophosphoesterase MPPED2; metallophosphoesterase MPPED2; metallophosphoesterase domain containing 2b; MPPED2; putative C11orf8 homolog; wu:fk34e08; zgc:56667
Gene Symbols MPPED2
Host Species Rabbit
Immunogen Recombinant Human Metallophosphoesterase MPPED2 protein (1-294AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 744, 77015
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.