Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mPR alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561376 Supplier Invitrogen™ Supplier No. PA561376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMAQKLSHL LPSLRQVIQE PQLSLQPEPV FTVDRAEVPP LFWKPYIYAG YRPLHQTWRF YFRTLFQQHN EA Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.

The steroid progesterone induces the resumption of maturation in oocytes via a nongenomic pathway through binding to a novel, membrane progestin receptor (mPR). This pathway inhibits adenylyl cyclase and reduces intracellular cAMP, and also activates mitogen-activated protein kinase to effect signal transduction pathways. Three distinct groups, designated α, β and γ, comprise this gene family. mPRα, also designated progestin and adipoQ receptor family member VII (PAQR7), consists of an extracellular N-terminus, an intracellular C-terminus, and seven transmembrane domains. It is expressed in ovary, testis, placenta, uterus and bladder. mPRβ, also designated progestin and adipoQ receptor family member VIII (PAQR8), consists of eight putative transmembrane regions and an intracellular N-terminus that contains a leucine-rich motif. It is a 354 amino acid protein with a molecular mass of about 40 kDa and is expressed in brain and spinal cord. Both mPRα and mPRβ may be G protein-coupled receptors and may be involved in oocyte maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mPR alpha
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAQR7
Gene Accession No. Q86WK9
Gene Alias 2310021M12Rik; AV001994; BB007237; Membrane progesterone P4 receptor alpha; Membrane progesterone receptor alpha; Membrane progestin receptor alpha; mPR; mPR alpha; Mpra; MRPA; mSR; PAQR7; Pglp; PPAR-gamma-induced liver protein; Progesterone and adipoQ receptor family member 7; Progestin and adipoQ receptor family member 7; progestin and adipoQ receptor family member VII
Gene Symbols PAQR7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human mPR alpha. Recombinant protein control fragment (Product #RP-99833).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.