Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ mPR alpha Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102102
Description
Antibody detects endogenous levels of total MPRA.
The steroid progesterone induces the resumption of maturation in oocytes via a nongenomic pathway through binding to a novel, membrane progestin receptor (mPR). This pathway inhibits adenylyl cyclase and reduces intracellular cAMP, and also activates mitogen-activated protein kinase to effect signal transduction pathways. Three distinct groups, designated α, β and γ, comprise this gene family. mPRα, also designated progestin and adipoQ receptor family member VII (PAQR7), consists of an extracellular N-terminus, an intracellular C-terminus, and seven transmembrane domains. It is expressed in ovary, testis, placenta, uterus and bladder. mPRβ, also designated progestin and adipoQ receptor family member VIII (PAQR8), consists of eight putative transmembrane regions and an intracellular N-terminus that contains a leucine-rich motif. It is a 354 amino acid protein with a molecular mass of about 40 kDa and is expressed in brain and spinal cord. Both mPRα and mPRβ may be G protein-coupled receptors and may be involved in oocyte maturation.
Specifications
mPR alpha | |
Polyclonal | |
Unconjugated | |
PAQR7 | |
2310021M12Rik; AV001994; BB007237; Membrane progesterone P4 receptor alpha; Membrane progesterone receptor alpha; Membrane progestin receptor alpha; mPR; mPR alpha; Mpra; MRPA; mSR; PAQR7; Pglp; PPAR-gamma-induced liver protein; Progesterone and adipoQ receptor family member 7; Progestin and adipoQ receptor family member 7; progestin and adipoQ receptor family member VII | |
Rabbit | |
Affinity chromatography | |
RUO | |
164091, 71904 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80ZE4, Q86WK9 | |
PAQR7 | |
A synthesized peptide derived from human PAQR7(Accession Q86WK9), corresponding to amino acid residues H313-K346. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction