Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mPR gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113495
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113495 Supplier Invitrogen™ Supplier No. PA5113495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mPR gamma Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

The steroid progesterone induces the resumption of maturation in oocytes via a nongenomic pathway through binding to a novel, membrane progestin receptor (mPR). This pathway inhibits adenylyl cyclase and reduces intracellular cAMP, and also activates mitogen-activated protein kinase to effect signal transduction pathways. Three distinct groups, designated alpha, beta, and gamma, comprise this gene family. While all contain 7 transmembrane domains, they show distinct distributions in reproductive, neural, kidney and intestinal tissues. These characteristics separate them from nuclear progestin receptors, and instead imply similarity to G-protein coupled receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mPR gamma
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PAQR5
Gene Accession No. Q9NXK6
Gene Alias 0610010I15Rik; AV002411; BB115488; Membrane progesterone P4 receptor gamma; Membrane progesterone receptor gamma; Membrane progestin receptor gamma; mPR gamma; Mprg; PAQR5; Pmrgamma; Progesterone and adipoQ receptor family member 5; Progestin and adipoQ receptor family member 5; progestin and adipoQ receptor family member V; progestin membrane receptor gamma; RGD1311259
Gene Symbols PAQR5
Host Species Rabbit
Immunogen Recombinant Human Membrane progestin receptor gamma protein (1-51AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54852
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.