Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPRIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554631 Supplier Invitrogen™ Supplier No. PA554631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDAYELEVLL RVKESEIQYL KQEISSLKDE LQTALRDKKY ASDKYKDIYT ELSIAKAKAD CDISRLKEQL KAATEALGEK SPDSATVSGY DIMKSKSNPD FLKKDRSCVT RQL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Targets myosin phosphatase to the actin cytoskeleton. Required for the regulation of the actin cytoskeleton by RhoA and ROCK1. Depletion leads to an increased number of stress fibers in smooth muscle cells through stabilization of actin fibers by phosphorylated myosin. Overexpression of MRIP as well as its F-actin-binding region leads to disassembly of stress fibers in neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPRIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MPRIP
Gene Accession No. P97434, Q6WCQ1, Q9ERE6
Gene Alias 9530046C02; AA536749; AI647711; C76423; KIAA0864; mKIAA0864; Mprip; Mrip; M-RIP; myosin phosphatase Rho interacting protein; myosin phosphatase Rho-interacting protein; myosin phosphatase-Rho interacting protein; p116 Rho interacting protein; p116Rip; Rho interacting protein 3; rho-interacting protein 3; Rho-interacting protein 3 (p116RIP) (RIP3); RHOIP3; RIP3
Gene Symbols MPRIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPRIP. Recombinant protein control fragment (Product #RP-93318).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116504, 23164, 26936
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.