Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Mpro Tracer
GREENER_CHOICESDP

Catalog No. 812650U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-265-0U 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 81-265-0U Supplier Tocris Bioscience™ Supplier No. 8126/50U
Add to Cart
Add to Cart
This item is not returnable. View return policy

SARS-CoV-2 main protease fluorescent ligand; TR-FRET acceptor

Mpro Tracer is a high affinity (Kd = 40 nM) non-covalent SARS-CoV-2 main protease fluorescent ligand. Mpro Tracer can be used in fluorescent polarisation (FP) assay for inhibitor screening/characterisation. It can be used as a FRET acceptor in time-resolved fluorescence resonance energy-transfer (TR-FRET) experiments. Excitation and emission maxima (λ) are 492 nm and 518 nm.

Specifications

Quantity 50 μg
Target Fluorescent Ligands
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.