Learn More
Invitrogen™ MPS1 Monoclonal Antibody (4A12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515522
Description
MA5-15522 targets MPS1 in indirect ELISA, IHC applications and shows reactivity with Human samples. The MA5-15522 immunogen is purified recombinant fragment of MPS1 expressed in E. Coli. MA5-15522 detects MPS1 which has a predicted molecular weight of approximately 95kDa.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
MPS1 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P42677 | |
RPS27 | |
Purified recombinant fragment of MPS1 expressed in E. Coli. | |
RUO | |
6232 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
4A12 | |
Unconjugated | |
RPS27 | |
3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.