Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588156 Supplier Invitrogen™ Supplier No. PA588156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVSAQWVAEA LRAPRAGQPL QLLDASWYLP KLGRDARREF EERHIPGAAF FDIDQCSDRT SPYDHMLPGA EHFAEYAGRL GVGAATHVVI YDASDQGLYS APRVWWMFRA FGHHAVSLLD GGLRHWLRQN LPLSSGKSQP A; Positive Samples: Mouse liver, Rat liver; Cellular Location: Cell junction, Cytoplasm, Mitochondrion, synapse, synaptosome.

This protein encoded by this gene catalyzes the transfer of a sulfur ion from 3-mercaptopyruvate to cyanide or other thiol compounds. It may be involved in cysteine degradation and cyanide detoxification. There is confusion in literature between this protein (mercaptopyruvate sulfurtransferase, MPST), which appears to be cytoplasmic, and thiosulfate sulfurtransferase (rhodanese, TST, GeneID:7263), which is a mitochondrial protein. Deficiency in MPST activity has been implicated in a rare inheritable disorder known as mercaptolactate-cysteine disulfiduria (MCDU). Alternatively spliced transcript variants encoding same or different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPST
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mpst
Gene Accession No. P97532, Q99J99
Gene Alias 3-mercaptopyruvate sulfurtransferase; human liver rhodanese; mercaptopyruvate sulfurtransferase; MPST; MST; testicular tissue protein Li 200; tRNA thiouridin modification protein 1; TST2; TUM1
Gene Symbols Mpst
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-170 of human MPST (NP_0669492).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192172, 246221
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.