Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPST Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $414.50
Specifications
Antigen | MPST |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145299
|
Invitrogen™
PA5145299 |
200 μL |
Each for $414.50
|
|
|||||
FEPA5145300
|
Invitrogen™
PA5145300 |
20 μL |
Each for $191.80
|
|
|||||
Description
This protein encoded by this gene catalyzes the transfer of a sulfur ion from 3-mercaptopyruvate to cyanide or other thiol compounds. It may be involved in cysteine degradation and cyanide detoxification. There is confusion in literature between this protein (mercaptopyruvate sulfurtransferase, MPST), which appears to be cytoplasmic, and thiosulfate sulfurtransferase (rhodanese, TST, GeneID:7263), which is a mitochondrial protein. Deficiency in MPST activity has been implicated in a rare inheritable disorder known as mercaptolactate-cysteine disulfiduria (MCDU). Alternatively spliced transcript variants encoding same or different isoforms have been identified for this gene.Specifications
MPST | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
P97532, Q99J99 | |
192172, 246221 | |
Recombinant Mouse MST protein, Met1-Gln297 (Accession #Q99J99). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
3-mercaptopyruvate sulfurtransferase; human liver rhodanese; mercaptopyruvate sulfurtransferase; MPST; MST; testicular tissue protein Li 200; tRNA thiouridin modification protein 1; TST2; TUM1 | |
Mpst | |
IgG | |
Protein A | |
Mpst |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title