Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPV17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100021
Description
Antibody detects endogenous levels of total MPV17.
This gene encodes a mitochondrial inner membrane protein that is implicated in the metabolism of reactive oxygen species. Mutations in this gene have been associated with the hepatocerebral form of mitochondrial DNA depletion syndrome.
Specifications
MPV17 | |
Polyclonal | |
Unconjugated | |
MPV17 | |
hMPV; h-MPV; Mpv17; Mpv-17; MpV17 mitochondrial inner membrane protein; MPV17 mitochondrial membrane protein-like; Mpv17 protein; Mpv17 transgene, kidney disease mutant-like; Mpv17, human homolog of glomerulosclerosis and nephrotic syndrome; MPV17, mitochondrial inner membrane protein; Mpv17l; MTDPS6; OTTHUMP00000122591; Protein Mpv17; SYM1; Tg.Mpv17 | |
Rabbit | |
Affinity chromatography | |
RUO | |
17527, 360463, 4358 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P19258, P39210, Q5BK62 | |
MPV17 | |
A synthesized peptide derived from human MPV17(Accession P39210), corresponding to amino acid residues V72-K122. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction