Learn More
Invitrogen™ MPV17 Polyclonal Antibody

Description
MPV17 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | MPV17 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | MPV17 |
| Gene Accession No. | P19258, P39210 |
| Gene Alias | hMPV; h-MPV; Mpv17; Mpv-17; MpV17 mitochondrial inner membrane protein; MPV17 mitochondrial membrane protein-like; Mpv17 protein; Mpv17 transgene, kidney disease mutant-like; Mpv17, human homolog of glomerulosclerosis and nephrotic syndrome; MPV17, mitochondrial inner membrane protein; Mpv17l; MTDPS6; OTTHUMP00000122591; Protein Mpv17; SYM1; Tg.Mpv17 |
| Gene Symbols | MPV17 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.