Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPV17L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559471 Supplier Invitrogen™ Supplier No. PA559471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQQSGDGTFK SAFTILYTKG TSATEGYPKK Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 40%.

Isoform 1 participates in reactive oxygen species metabolism by up- or down-regulation of the genes of antioxidant enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPV17L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mpv17l
Gene Accession No. Q2QL34
Gene Alias M-LP; M-LP homolog; M-LPH; MLPH1; MLPH2; MPV17 mitochondrial inner membrane protein like; MPV17 mitochondrial membrane protein-like; Mpv17 transgene, kidney disease mutant-like; MPV17L; MPV17L1; mpv17-like protein; Mpv17-like protein type 1; Mpv17-like protein type 2
Gene Symbols Mpv17l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPV17L. Recombinant protein control fragment (Product #RP-98687).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.