Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPV17L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570139 Supplier Invitrogen™ Supplier No. PA570139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 100%; Human: 100%.

Isoform 1 participates in reactive oxygen species metabolism by up- or down-regulation of the genes of antioxidant enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPV17L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mpv17l
Gene Accession No. Q2QL34
Gene Alias M-LP; M-LP homolog; M-LPH; MLPH1; MLPH2; MPV17 mitochondrial inner membrane protein like; MPV17 mitochondrial membrane protein-like; Mpv17 transgene, kidney disease mutant-like; MPV17L; MPV17L1; mpv17-like protein; Mpv17-like protein type 1; Mpv17-like protein type 2
Gene Symbols Mpv17l
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human MPV17L.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255027
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.