Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPZL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588952 Supplier Thermo Scientific Supplier No. PA588952
Add to Cart
Add to Cart

MPZL1 Polyclonal Antibody

MPZL1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MPZL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mpzl1
Gene Accession No. O95297, Q3TEW6, Q6AYT8
Gene Alias 1110007A10Rik; FLJ21047; immunoglobulin family transmembrane protein; Mpzl1; MPZL1b; myelin protein zero like 1; myelin protein zero-like 1; myelin protein zero-like protein 1; protein zero related; protein zero-related; Pzr; PZR1b; PZRa; PZRb; UNQ849/PRO1787
Gene Symbols Mpzl1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 195-269 of human MPZL1 (NP_0039441).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360871, 68481, 9019
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.