Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPZL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115812 Supplier Invitrogen™ Supplier No. PA5115812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PZR.

Cell surface receptor, which is involved in signal transduction processes. Recruits PTPN11/SHP-2 to the cell membrane and is a putative substrate of PTPN11/SHP-2. Is a major receptor for concanavalin-A (ConA) and is involved in cellular signaling induced by ConA, which probably includes Src family tyrosine-protein kinases. Isoform 3 seems to have a dominant negative role; it blocks tyrosine phosphorylation of MPZL1 induced by ConA. Isoform 1, but not isoform 2 and isoform 3, may be involved in regulation of integrin-mediated cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPZL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Mpzl1
Gene Accession No. O95297, Q3TEW6
Gene Alias 1110007A10Rik; FLJ21047; immunoglobulin family transmembrane protein; Mpzl1; MPZL1b; myelin protein zero like 1; myelin protein zero-like 1; myelin protein zero-like protein 1; protein zero related; protein zero-related; Pzr; PZR1b; PZRa; PZRb; UNQ849/PRO1787
Gene Symbols Mpzl1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MPZL1(Accession O95297), corresponding to amino acid residues H235-N269.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68481, 9019
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.