Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPZL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734940 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4940 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4940 Supplier Proteintech Group Inc Supplier No. 255131AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This polyclonal antibody raised against 32-235 aa of human MPZL3 reveals bands of 28 kDa and 70 kDa, which could be endogenous MPZL3 of unmodified form and post-translational modified form, respectively.

MPZL3 mediates homophilic cell-cell adhesion.

Specifications

Antigen MPZL3
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MPZL3
Gene Accession No. Q3V3F6, Q6UWV2
Gene Alias MPZL3, myelin protein zero like 3
Gene Symbols Mpzl3
Host Species Rabbit
Immunogen MPZL3 Fusion Protein Ag21905
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196264, 319742
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.