Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MR1 Mouse anti-Human, Mouse, Alexa Fluor™ 647, Clone: 8F2.F9, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568669
Description
MR1 Monoclonal antibody specifically detects MR1 in Human|Mouse samples. It is validated for Flow cytometry
Specifications
MR1 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
8F2.F9 | |
Alexa Fluor 647 | |
HLALS; MHC class I-like antigen MR-1; MHC class-I related-gene protein | |
MR1 Extracellular Domain Complexed with β2m | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction