Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MR1 Mouse anti-Human, Mouse, PE, Clone: 8F2.F9, BD Pharmingen™

Catalog No. BDB568013 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568013 25 μg
BDB568012 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568013 Supplier BD Biosciences Supplier No. 568013
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MR1 Monoclonal antibody specifically detects MR1 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MR1
Applications Flow Cytometry
Classification Monoclonal
Clone 8F2.F9
Concentration 0.2 mg/mL
Conjugate PE
Dilution 0.02% Sodium Azide
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias HLALS; MHC class I-like antigen MR-1; MHC class-I related-gene protein
Host Species Mouse
Immunogen MR1 Extracellular Domain Complexed with β2m
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.