Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596909 Supplier Invitrogen™ Supplier No. PA596909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse skeletal muscle; Cellular Location: Cell membrane, Endoplasmic reticulum, Endoplasmic reticulum membrane, Extracellular side, Single-pass membrane protein, Single-pass type I membrane protein.

Antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. Involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain called mucosal-associated invariant T- cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and therefore may be involved in monitoring commensal flora or serve as a distress signal. Expression and MAIT cell recognition seem to be ligand-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MR1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MR1
Gene Accession No. O19477, Q8HWB0, Q95460
Gene Alias Class I histocompatibility antigen-like protein; H2ls; histocompatibility-2 complex class 1-like sequence; Hlals; Major histocompatibility complex class I-related gene protein; major histocompatibility complex, class I-like sequence; major histocompatibility complex, class I-related; MCH class I; MHC class I-like antigen MR-1; MHC class I-like sequence; MHC class I-related gene protein; MHC class-I related-gene protein; MR1; Mr1a
Gene Symbols MR1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15064, 25119, 3140
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.