Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MR1 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8609
![]()
|
Proteintech Group Inc
132601AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. Involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain called mucosal-associated invariant T- cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and therefore may be involved in monitoring commensal flora or serve as a distress signal. Expression and MAIT cell recognition seem to be ligand-dependent.Specifications
| MR1 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O19477, Q8HWB0, Q95460 | |
| 15064, 25119, 3140 | |
| MR1 Fusion Protein Ag3849 | |
| Primary | |
| -20°C | |
| MR1 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HLALS, MR1 | |
| Mr1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title