Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571793
Description
MRAP2 Polyclonal Antibody for Western Blot, IHC (P)
Modulator of melanocortin receptor 4 (MC4R), a receptor involved in energy homeostasis. Plays a central role in the control of energy homeostasis and body weight regulation by increasing ligand-sensitivity of MC4R and MC4R-mediated generation of cAMP. May also act as a negative regulator of MC2R: competes with MRAP for binding to MC2R and impairs the binding of corticotropin (ACTH) to MC2R. May also regulate activity of other melanocortin receptors (MC1R, MC3R and MC5R); however, additional evidences are required in vivo.
Specifications
MRAP2 | |
Polyclonal | |
Unconjugated | |
MRAP2 | |
bA51G5.2; BB633055; BMIQ18; C6orf117; MC2R accessory protein 2; melanocortin 2 receptor accessory protein 2; melanocortin-2 receptor accessory protein 2; Mrap2; RGD1309873 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
112609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.87 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q96G30 | |
MRAP2 | |
KLH conjugated synthetic peptide between 58-86 amino acids from the Central region of human MRAP2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction