Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113283
Description
MRAP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Modulator of melanocortin receptor 4 (MC4R), a receptor involved in energy homeostasis. Plays a central role in the control of energy homeostasis and body weight regulation by increasing ligand-sensitivity of MC4R and MC4R-mediated generation of cAMP. May also act as a negative regulator of MC2R: competes with MRAP for binding to MC2R and impairs the binding of corticotropin (ACTH) to MC2R. May also regulate activity of other melanocortin receptors (MC1R, MC3R and MC5R); however, additional evidences are required in vivo.
Specifications
MRAP2 | |
Polyclonal | |
Unconjugated | |
MRAP2 | |
bA51G5.2; BB633055; BMIQ18; C6orf117; MC2R accessory protein 2; melanocortin 2 receptor accessory protein 2; melanocortin-2 receptor accessory protein 2; Mrap2; RGD1309873 | |
Rabbit | |
Protein G | |
RUO | |
112609, 244958 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
D3Z1Q2, Q96G30 | |
MRAP2 | |
Recombinant Human Melanocortin-2 receptor accessory protein 2 protein (66-205AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction