Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571377 Supplier Invitrogen™ Supplier No. PA571377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRAS Polyclonal Antibody for Western Blot

MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRAS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRAS
Gene Accession No. O08989
Gene Alias 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras
Gene Symbols MRAS
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human MRAS.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17532
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.