Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRBG Indicator Methyl Red-Bromocresol Green, Reagents
Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | MRBG Indicator Methyl Red-Bromocresol Green |
Boiling Point | 72°C |
CAS | 64-17-5, 62625-32-5, 845-10-3 |
Synonym | Methyl Red - Bromocresol Green |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 0.791 |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction