Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MRC1 Recombinant Protein

Catalog No. 89968773
Click to view available options
:
25 μg

Used for AP, Array, ELISA, WB-Re

Sequence: TRQFLIYNEDHKRCVDAVSPSAVQTAACNQDAESQKFRWVSESQIMSVAFKLCLGVPSKTDWVAITLYACDSKSEFQKWECKNDTLLGIKGEDLFFNYGNRQEKNIMLY

Specifications

Accession Number NP_002429
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH = 8.0 in the elution buffer.
Gene ID (Entrez) 4360
Molecular Weight (g/mol) 37.73 kDa
Name MRC1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Immunogen TRQFLIYNEDHKRCVDAVSPSAVQTAACNQDAESQKFRWVSESQIMSVAFKLCLGVPSKTDWVAITLYACDSKSEFQKWECKNDTLLGIKGEDLFFNYGNRQEKNIMLY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD206, CLEC13D, MMR
Common Name MRC1
Gene Symbol MRC1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System Wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.