Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mrc2 Antibody [Janelia Fluor™ 646], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4789J
Description
Mrc2 Polyclonal antibody specifically detects Mrc2 in Mouse samples. It is validated for Western BlotSpecifications
Mrc2 | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
Mouse myeloma cell line NS0-derived recombinant mouse Mrc2, Ala31-Ala1413, Accession # AAI16643 | |
0.1 mL | |
CD Markers, Cell Biology, Immunology | |
9902 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction