Learn More
Invitrogen™ MRC2 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
$459.00 - $1159.00
Specifications
Antigen | MRC2 Biosimilar |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
Classification | Recombinant Monoclonal |
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Quark patent anti-ENDO 180.
May play a role as endocytotic lectin receptor displaying calcium-dependent lectin activity. Internalizes glycosylated ligands from the extracellular space for release in an endosomal compartment via clathrin-mediated endocytosis. May be involved in plasminogen activation system controlling the extracellular level of PLAUR/PLAU, and thus may regulate protease activity at the cell surface. May contribute to cellular uptake, remodeling and degradation of extracellular collagen matrices. May play a role during cancer progression as well as in other chronic tissue destructive diseases acting on collagen turnover. May participate in remodeling of extracellular matrix cooperating with the matrix metalloproteinases (MMPs).Specifications
MRC2 Biosimilar | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Lyophilized | |
IgG1 | |
Human | |
Primary | |
Protein A |
1 mg/mL | |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
Unconjugated | |
Human | |
RUO | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.