Learn More
Invitrogen™ MRC2 Polyclonal Antibody

Description
MRC2 Polyclonal Antibody detects endogenous levels of MRC2 protein.
Specifications
Specifications
| Antigen | MRC2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | MRC2 |
| Gene Accession No. | Q4TU93, Q64449, Q9UBG0 |
| Gene Alias | CD280; CLEC13E; collagen-binding factor Endo180; C-type lectin domain family 13 member E; C-type mannose receptor 2; Endo180; endocytic receptor (macrophage mannose receptor family); endocytic receptor 180; KIAA0709; Lectin lambda; macrophage mannose receptor 2; mannose receptor C type 2; mannose receptor, C type 2; mKIAA0709; Mrc2; novel lectin; UPARAP; UPAR-associated protein; urokinase-type plasminogen activator receptor-associated protein |
| Gene Symbols | MRC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.