Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRCK Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146822H
Description
MRCK Polyclonal antibody specifically detects MRCK in Human samples. It is validated for Western BlotSpecifications
MRCK | |
Polyclonal | |
Western Blot | |
CDC42 binding protein kinase alpha (DMPK-like), CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, EC 2.7.11, EC 2.7.11.1, KIAA0451DKFZp686L1738, MRCK, MRCK alpha, MRCKADKFZp686P1738, Myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, Myotonic dystrophy protein kinase-like alpha, PK428FLJ23347, serine/threonine-protein kinase MRCK alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase | |
The immunogen for this product maps to a region between residue 1560 and 1610 of human myotonic dystrophy protein kinase-like alpha using the numbering given in entry CAH71185.1 (GeneID 8476). | |
0.1 mL | |
Protein Kinase | |
8476 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction