Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRCK gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100763 Supplier Invitrogen™ Supplier No. PA5100763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRCKG.

CDC42 BPG serine/threonine kinase is encoded by the CDC42BPG gene. It acts as a downstream effector of CDC42 in cytoskeletal reorganization, and contributes to the actomyosin contractility required for cell invasion, through the regulation of MYPT1 and MLC2 phosphorylation. It belongs to the AGC protein kinase family, DMPK subfamily. It contains an N-terminal kinase domain, followed by a coiled-coil domain, a second coiled-coil domain containing a kinase inhibitory motif (KIM), a variable region, a cysteine-rich domain, a pleckstrin (PLEK; 173570) homology (PH) domain, a citron (605629) homology (CNH) domain, and a C-terminal CDC42 (116952)/RAC (602048) interactive binding (CRIB) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRCK gamma
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDC42BPG
Gene Accession No. Q6DT37, Q80UW5
Gene Alias BC046418; CDC42 binding protein kinase gamma; CDC42 binding protein kinase gamma (DMPK-like); CDC42-binding protein kinase gamma; CDC42BPG; DMPK2; DMPK-like gamma; HSMDPKIN; KAPPA-200; MRCK gamma; MRCKG; MRCKgamma; Myotonic dystrophy kinase-related CDC42-binding kinase gamma; myotonic dystrophy protein kinase like protein; myotonic dystrophy protein kinase-like alpha; myotonic dystrophy protein kinase-like gamma; serine/threonine-protein kinase MRCK gamma
Gene Symbols CDC42BPG
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDC42BPG(Accession Q6DT37), corresponding to amino acid residues L1393-F1443.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240505, 55561
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.