Learn More
MRCKalpha Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302693AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
MRCK alpha (myotonic dystrophy kinase-related Cdc42-binding kinase alpha) is a member of the dystrophia myotonica protein kinase (DMPK) family that functions downstream of Cdc42 in actin cytoskeletal reorganization. It is a serine/threonine kinase with multiple functional domains, including phorbol ester-responsive C1 domain. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.Specifications
MRCKalpha | |
Polyclonal | |
Unconjugated | |
CDC42BPA | |
RP5-1087E8.4, DKFZp686L1738, DKFZp686P1738, FLJ23347, KIAA0451, MRCK, MRCKA, PK428, CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, MRCK alpha, myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, myotonic dystrophy protein kinase-like alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase, serine/threonine-protei | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8476 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q5VT25 | |
CDC42BPA | |
Between 1560 and 1610 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.