Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRCKalpha Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563236
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3236 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3236 Supplier Bethyl Laboratories, Inc Supplier No. A302694AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

MRCK alpha (myotonic dystrophy kinase-related Cdc42-binding kinase alpha) is a member of the dystrophia myotonica protein kinase (DMPK) family that functions downstream of Cdc42 in actin cytoskeletal reorganization. It is a serine/threonine kinase with multiple functional domains, including phorbol ester-responsive C1 domain. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.

Specifications

Antigen MRCKalpha
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CDC42BPA
Gene Accession No. Q5VT25
Gene Alias RP5-1087E8.4, DKFZp686L1738, DKFZp686P1738, FLJ23347, KIAA0451, MRCK, MRCKA, PK428, CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, MRCK alpha, myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, myotonic dystrophy protein kinase-like alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase, serine/threonine-protei
Gene Symbols CDC42BPA
Host Species Rabbit
Immunogen Between 1610 and 1660
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8476
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.