Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mre11 Antibody (15B8.1E7.6), Allophycocyanin, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP259677APC
Description
Mre11 Monoclonal specifically detects Mre11 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
Mre11 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
MRE11 | |
Mre11 Antibody (15B8.1E7.6) [Allophycocyanin] was made to a partial recombinant mouse Mre11 protein (amino acids 68-608) [UniProt Q61216] | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
4361 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
15B8.1E7.6 | |
Flow Cytometry, Flow (Intracellular) | |
ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction