Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRE11 Armenian Hamster anti-Human, Mouse, MilliporeSigma™

Catalog No. MABE115325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1153-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1153-25 Supplier MilliporeSigma Supplier No. MABE115325UG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Anti-MRE11, clone 15B8.1E7.6 is a highly specific armenian hamster monoclonal antibody, that targets Double-strand break repair protein MRE11A and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRE11
Applications Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Mre11a,Mre11
Host Species Armenian Hamster
Immunogen Recombinant fragment corresponding to 540 amino acids from murine Double-strand break repair protein MRE11A.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 15B8.1E7.6 detects Double-strand break repair protein MRE11A in human and murine cells.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.