Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC29414 and based on accession number NM_5591. The protein contains a C-terminal DDK tag. |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens MRE11 meiotic recombination 11 homolog A (S. cerevisiae) (MRE11A), transcript variant 1, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Research Category | Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase |
| Synonym | ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B |
| Protein State | Native |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
