Learn More
Invitrogen™ MRE11 Polyclonal Antibody

Description
A suggested positive control is rat lung tissue lysate. PA5-20870 can be used with blocking peptide PEP-0984.
Specifications
Specifications
| Antigen | MRE11 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Mre11 |
| Gene Accession No. | P49959, Q61216, Q9JIM0 |
| Gene Alias | ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C |
| Gene Symbols | Mre11, Mre11a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.