Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRE11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586361 Supplier Invitrogen™ Supplier No. PA586361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

MRE11 (double-strand break repair protein MRE11) is a component of the MRN complex. This complex plays a central role in double-strand break repair, DNA recombination, maintenance of telomere integrity and meiosis. The complex possesses single-stranded endonuclease activity and double-strand-specific 3'-5' exonuclease activity which are provided by MRE11. RAD50 is required to bind DNA ends and hold them in close proximity. Mutations in the gene could result in ataxia-telangiectasia-like disorder 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRE11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Mre11
Gene Accession No. P49959, Q61216, Q9JIM0
Gene Alias ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C
Gene Symbols Mre11, Mre11a
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 240-280 of Human MRE11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17535, 4361, 64046
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.