Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mre11 Rabbit anti-Human, Mouse, Clone: 8Q2B3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Mre11 |
---|---|
Clone | 8Q2B3 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Mre11 Monoclonal antibody specifically detects Mre11 in Human, Mouse samples. It is validated for Western BlotSpecifications
Mre11 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Mre11 (P49959). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8Q2B3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4361 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title