Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRFAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566011 Supplier Invitrogen™ Supplier No. PA566011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNHLQNPGDAA EGRAAKRCEK Highest antigen sequence identity to the following orthologs - mouse 43%, rat 43%.

This gene encodes an intracellular protein that interacts with members of the MORF4/MRG (mortality factor on chromosome 4/MORF4 related gene) family and the tumor suppressor Rb (retinoblastoma protein.) The protein may play a role in senescence, cell growth and immortalization. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRFAP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRFAP1
Gene Accession No. Q9Y605
Gene Alias 9130413I22Rik; AL022827; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; Morf4 family associated protein 1 like 1; Morf4 family associated protein 1-like 1; MORF4 family-associated protein 1; MORF4 family-associated protein 1-like 1; MRFAP1; MRFAP1L1; PAM14; PGR1; PP784; protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; Protein PGR1; T-cell activation protein
Gene Symbols MRFAP1
Host Species Rabbit
Immunogen Recombinant Human MRFAP1. Recombinant protein control fragment (Product #RP-106695).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.