Learn More
Invitrogen™ MRFAP1L1 Polyclonal Antibody

Description
MRFAP1L1 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | MRFAP1L1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MRFAP1L1 |
| Gene Accession No. | Q96HT8 |
| Gene Alias | Morf4 family associated protein 1 like 1; Morf4 family associated protein 1-like 1; MORF4 family-associated protein 1-like 1; MRFAP1L1; PP784 |
| Gene Symbols | MRFAP1L1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.