Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRFAP1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112614
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112614 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112614 Supplier Invitrogen™ Supplier No. PA5112614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRFAP1L1 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRFAP1L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRFAP1L1
Gene Accession No. Q96HT8
Gene Alias Morf4 family associated protein 1 like 1; Morf4 family associated protein 1-like 1; MORF4 family-associated protein 1-like 1; MRFAP1L1; PP784
Gene Symbols MRFAP1L1
Host Species Rabbit
Immunogen Recombinant Human MORF4 family-associated protein 1-like 1 protein (1-127AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114932
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.